Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ggnbp2
Entrez Gene ID
217039
Gene Name
gametogenetin binding protein 2
Gene Aliases
AI451896, C75939, D330017P12Rik, DIF-3, Zfp403
Chromosome Location
Chr.11: 84832361 - 84870738 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153144.2 NP_694784.2 10 - 11 1577 108
GenBank mRNA AB064543.1 - 11 - 12 1706 108
AY633741.1 - 10 - 11 1577 108
BC139772.1 - 9 - 10 1477 108
BC145605.1 - 10 - 11 1605 108
BC150801.1 - 10 - 11 1605 108

View Full Product Details