The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029934.1 | NP_001025105.1 | 12 - 13 | 1430 | 104 |
GenBank mRNA | AK148221.1 | - | 12 - 13 | 1430 | 104 |