The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019816.1 | NP_062790.1 | 12 - 13 | 1632 | 58 |
GenBank mRNA | AF222801.1 | - | 12 - 13 | 1632 | 58 |
AK016820.1 | - | 8 - 9 | 869 | 58 | |
AK141334.1 | - | 12 - 13 | 1630 | 58 | |
BC025080.1 | - | 12 - 13 | 1619 | 58 |