Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nkap
Entrez Gene ID
67050
Gene Name
NFKB activating protein
Gene Aliases
2610020O08Rik, AA987160, AI849147, AL024236
Chromosome Location
Chr.X: 37126763 - 37150746 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025937.4 NP_080213.3 6 - 7 1077 92
GenBank mRNA AK011492.1 - 6 - 7 1028 92
AK089943.1 - 6 - 7 1005 92
AY388959.1 - 6 - 7 1028 92
BC026774.1 - 6 - 7 1045 92

View Full Product Details