The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_201406.1 | NP_958808.1 | 6 - 7 | 731 | 59 |
GenBank mRNA | AK033331.1 | - | 6 - 7 | 705 | 59 |
AK039604.1 | - | 6 - 7 | 702 | 59 | |
AK043232.1 | - | 6 - 7 | 705 | 59 | |
AK045759.1 | - | 6 - 7 | 702 | 59 | |
AK053771.1 | - | 6 - 7 | 705 | 59 | |
AK142871.1 | - | 6 - 7 | 702 | 59 | |
AK151858.1 | - | 6 - 7 | 693 | 59 | |
AK152229.1 | - | 6 - 7 | 693 | 59 | |
AK153300.1 | - | 6 - 7 | 697 | 59 | |
AK166408.1 | - | 6 - 7 | 708 | 59 | |
AK169633.1 | - | 5 - 6 | 504 | 59 | |
BC058979.1 | - | 6 - 7 | 731 | 59 |