Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bzrap1
Entrez Gene ID
207777
Gene Name
benzodiazepine receptor associated protein 1
Gene Aliases
D230016K05, PRAX-1, mKIAA0612
Chromosome Location
Chr.11: 87759066 - 87785928 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172449.2 NP_766037.2 21 - 22 4722 88

View Full Product Details