The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172449.2 | NP_766037.2 | 8 - 9 | 2040 | 75 |
GenBank mRNA | CN460580.1 | - | 2 - 3 | 152 | 75 |