The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029856.2 | NP_001025027.1 | 20 - 21 | 4767 | 64 |
GenBank mRNA | AY557610.1 | - | 20 - 21 | 4758 | 64 |
BC046463.1 | - | 16 - 17 | 2064 | 64 | |
BC141180.1 | - | 19 - 20 | 4263 | 64 |