The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017470.2 | NP_059498.2 | 3 - 4 | 396 | 95 |
GenBank mRNA | AB010031.1 | - | 3 - 4 | 374 | 95 |
AK029618.1 | - | 2 - 3 | 207 | 95 | |
AK132061.1 | - | 3 - 4 | 386 | 95 | |
BC005426.1 | - | 3 - 4 | 325 | 95 |