The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175109.3 | NP_780318.1 | 2 - 3 | 216 | 58 |
GenBank mRNA | AB233218.1 | - | 2 - 3 | 212 | 58 |
AK010954.1 | - | 2 - 3 | 200 | 58 | |
AK075974.1 | - | 2 - 3 | 197 | 58 | |
BC038691.1 | - | 2 - 3 | 202 | 58 | |
BC050115.1 | - | 2 - 3 | 205 | 58 | |
BC065381.1 | - | 1 - 2 | 149 | 58 | |
BY237968.1 | - | 2 - 3 | 216 | 58 |