Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kpnb1
Entrez Gene ID
16211
Gene Name
karyopherin (importin) beta 1
Gene Aliases
AA409963, IPOB, Impnb
Chromosome Location
Chr.11: 97159710 - 97187892 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008379.3 NP_032405.3 10 - 11 1533 102
GenBank mRNA AK077709.1 - 10 - 11 1524 102
AK147175.1 - 10 - 11 1552 102
AK169171.1 - 10 - 11 1500 102
BC052438.1 - 10 - 11 1288 102
BC052711.1 - 10 - 11 1321 102
BC055115.1 - 10 - 11 1518 102
D45836.1 - 10 - 11 1424 102
D67015.1 - 10 - 11 1261 102

View Full Product Details