Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pnpla3
Entrez Gene ID
116939
Gene Name
patatin-like phospholipase domain containing 3
Gene Aliases
Adpn
Chromosome Location
Chr.15: 84167783 - 84189521 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_054088.3 NP_473429.2 5 - 6 819 74
GenBank mRNA AY037763.1 - 5 - 6 799 74
BC028792.1 - 5 - 6 807 74

View Full Product Details