The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018873.2 | NP_061361.2 | 2 - 3 | 462 | 114 |
GenBank mRNA | AF040944.1 | - | 2 - 3 | 467 | 114 |
U59873.1 | - | 2 - 3 | 466 | 114 |