The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085500.2 | NP_001078969.2 | 2 - 3 | 380 | 71 |
GenBank mRNA | AA987107.1 | - | 2 - 3 | 395 | 71 |
BC056217.1 | - | 2 - 3 | 375 | 71 | |
BC147739.1 | - | 3 - 4 | 475 | 71 | |
BC147742.1 | - | 3 - 4 | 475 | 71 | |
BC157903.1 | - | 3 - 4 | 418 | 71 |