Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cacna1i
Entrez Gene ID
239556
Gene Name
calcium channel, voltage-dependent, alpha 1I subunit
Gene Aliases
Ca(V)3.3
Chromosome Location
Chr.15: 80287238 - 80398292 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044308.2 NP_001037773.2 34 - 35 6156 146
GenBank mRNA AK147585.1 - 34 - 35 6156 146

View Full Product Details