The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145476.2 | NP_663451.2 | 12 - 13 | 1535 | 73 |
GenBank mRNA | AK039627.1 | - | 12 - 13 | 1548 | 73 |
AK146872.1 | - | 12 - 13 | 1536 | 73 | |
AK154226.1 | - | 12 - 13 | 1557 | 73 | |
AK154602.1 | - | 12 - 13 | 1578 | 73 | |
AK155459.1 | - | 12 - 13 | 1549 | 73 | |
AK155770.1 | - | 12 - 13 | 1578 | 73 | |
AK178967.1 | - | 2 - 3 | 114 | 73 | |
BC023106.1 | - | 9 - 10 | 930 | 73 | |
BC066009.1 | - | 12 - 13 | 1535 | 73 |