The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009888.3 | NP_034018.2 | 19 - 20 | 3333 | 94 |
GenBank mRNA | BC066092.1 | - | 19 - 20 | 3202 | 94 |
M12660.1 | - | 19 - 20 | 3244 | 94 |