Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cfh
Entrez Gene ID
12628
Gene Name
complement component factor h
Gene Aliases
Mud-1, NOM, Sas-1, Sas1
Chromosome Location
Chr.1: 140085855 - 140183411 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009888.3 NP_034018.2 1 - 2 244 136
GenBank mRNA AK052688.1 - 1 - 2 244 136
AK087707.1 - 1 - 2 153 136
BB661130.1 - 1 - 2 244 136
BC066092.1 - 1 - 2 113 136
M12660.1 - 1 - 2 155 136

View Full Product Details