Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Snx11
Entrez Gene ID
74479
Gene Name
sorting nexin 11
Gene Aliases
4933439F10Rik, A930041K09Rik, AI117694
Chromosome Location
Chr.11: 96767546 - 96777697 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163389.1 NP_001156861.1 1 - 2 96 118
NM_028965.4 NP_083241.1 1 - 2 96 118
GenBank mRNA AK037747.1 - 1 - 2 68 118
AK042409.1 - 1 - 2 72 118
AK076332.1 - 1 - 2 72 118

View Full Product Details