The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198295.2 | NP_938037.2 | 15 - 16 | 1285 | 112 |
GenBank mRNA | AK035946.1 | - | 15 - 16 | 1233 | 112 |
AK042787.1 | - | 15 - 16 | 1228 | 112 | |
AK129451.1 | - | 15 - 16 | 1179 | 112 | |
AK140709.1 | - | 15 - 16 | 1286 | 112 | |
BC057139.1 | - | 10 - 11 | 753 | 112 |