The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029447.2 | NP_083723.1 | 12 - 13 | 2136 | 85 |
GenBank mRNA | AK015561.1 | - | 1 - 2 | 145 | 85 |
AK050120.1 | - | 2 - 3 | 250 | 85 | |
AK079090.1 | - | 12 - 13 | 2137 | 85 | |
AK146364.1 | - | 12 - 13 | 2085 | 85 | |
AK170508.1 | - | 12 - 13 | 2137 | 85 | |
AK171432.1 | - | 12 - 13 | 2137 | 85 | |
BC016224.1 | - | 12 - 13 | 2109 | 85 | |
BC025520.1 | - | 5 - 6 | 995 | 85 |