The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016915.4 | NP_058611.1 | 1 - 2 | 290 | 82 |
GenBank mRNA | BC003487.1 | - | 1 - 2 | 83 | 82 |
BC052845.1 | - | 1 - 2 | 115 | 82 | |
BY099728.1 | - | 1 - 2 | 127 | 82 |