Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
Dnah8
Entrez Gene ID
13417
Gene Name
dynein, axonemal, heavy chain 8
Gene Aliases
ATPase, Dnahc8, Hst6.7b, P1-Loop
Chromosome Location
Chr.17: 30626936 - 30875762 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013811.3 NP_038839.2 - 2587 71
GenBank mRNA AF356520.1 - - 2587 71
AF356521.1 - - 2587 71
AF356522.1 - - 2597 71
AF356523.1 - - 2597 71
AF363577.1 - - 2597 71

View Full Product Details