The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177642.3 | NP_808310.2 | 17 - 18 | 6421 | 95 |
GenBank mRNA | EU429481.1 | - | 3 - 4 | 3293 | 95 |
EU429482.1 | - | 2 - 3 | 225 | 95 | |
EU429483.2 | - | 12 - 13 | 1574 | 95 | |
GU290042.1 | - | 17 - 18 | 6421 | 95 |