Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ing5
- Entrez Gene ID
-
66262
- Gene Name
- inhibitor of growth family, member 5
- Gene Aliases
-
1700001C14Rik, 1700027H23Rik, 1810018M11Rik, AI225768
- Chromosome Location
-
Chr.1: 93802051 - 93822101 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK006421.1 |
- |
1 - 2 |
179 |
100 |
View Full Product Details