Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Abca17
Entrez Gene ID
381072
Gene Name
ATP-binding cassette, sub-family A (ABC1), member 17
Gene Aliases
-
Chromosome Location
Chr.17: 24264258 - 24351897 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB112584.1 - 10 - 11 1579 91

View Full Product Details