Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tbx2
Entrez Gene ID
21385
Gene Name
T-box 2
Gene Aliases
-
Chromosome Location
Chr.11: 85832615 - 85841948 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009324.2 NP_033350.2 5 - 6 1550 73
GenBank mRNA AK137225.1 - 5 - 6 760 73
AK149348.1 - 5 - 6 1358 73
U15566.1 - 5 - 6 1086 73

View Full Product Details