The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033073.3 | NP_149064.1 | 2 - 3 | 592 | 94 |
GenBank mRNA | AF509888.1 | - | 2 - 3 | 569 | 94 |
AK002434.1 | - | 2 - 3 | 570 | 94 | |
AK137217.1 | - | - | 570 | 94 | |
AK146914.1 | - | 2 - 3 | 570 | 94 | |
BC030403.1 | - | 2 - 3 | 603 | 94 |