The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033073.3 | NP_149064.1 | 8 - 9 | 1296 | 69 |
GenBank mRNA | AF509888.1 | - | 8 - 9 | 1273 | 69 |
AK002434.1 | - | 8 - 9 | 1274 | 69 | |
AK137217.1 | - | - | 1274 | 69 | |
AK146914.1 | - | 8 - 9 | 1274 | 69 | |
BC010337.1 | - | 7 - 8 | 3290 | 69 | |
BC030403.1 | - | 8 - 9 | 1307 | 69 | |
BU755120.1 | - | 1 - 2 | 69 |