Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl30
Entrez Gene ID
70788
Gene Name
kelch-like 30
Gene Aliases
4631423F02Rik
Chromosome Location
Chr.1: 91351073 - 91362404 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027551.2 NP_081827.1 2 - 3 907 84
GenBank mRNA AK086025.1 - 2 - 3 908 84
AK146580.1 - 2 - 3 964 84
AK148398.1 - 2 - 3 915 84
AK182611.1 - 1 - 2 141 84
BC115428.1 - 1 - 2 850 84
BC115429.1 - 1 - 2 850 84

View Full Product Details