Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Traf7
Entrez Gene ID
224619
Gene Name
TNF receptor-associated factor 7
Gene Aliases
RFWD1
Chromosome Location
Chr.17: 24508850 - 24527938 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172113.1 NP_001165584.1 2 - 3 291 56
NM_001172114.1 NP_001165585.1 2 - 3 291 56
GenBank mRNA AK088431.1 - 2 - 3 200 56
AK131617.1 - 2 - 3 212 56
AK184420.1 - 1 - 2 35 56

View Full Product Details