The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172113.1 | NP_001165584.1 | 4 - 5 | 437 | 57 |
NM_153792.2 | NP_722487.2 | 4 - 5 | 457 | 57 | |
GenBank mRNA | BC008598.1 | - | 4 - 5 | 360 | 57 |