The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040187.1 | NP_001035277.1 | 11 - 12 | 1707 | 103 |
NM_199080.2 | NP_951062.1 | 11 - 12 | 1707 | 103 | |
GenBank mRNA | AK151061.1 | - | 11 - 12 | 1482 | 103 |
AK152749.1 | - | 11 - 12 | 1483 | 103 | |
AK152831.1 | - | 11 - 12 | 1482 | 103 | |
BC096036.1 | - | 11 - 12 | 1707 | 103 | |
BI655302.1 | - | 4 - 5 | 379 | 103 |