Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eif3l
Entrez Gene ID
223691
Gene Name
eukaryotic translation initiation factor 3, subunit L
Gene Aliases
0610011H21Rik, D15N1e, Eif3eip, Eif3ip, Eif3s6ip, HSP-66Y, PAF67
Chromosome Location
Chr.15: 79075223 - 79094400 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145139.2 NP_660121.2 1 - 2 69 59
GenBank mRNA AJ310346.1 - 1 - 2 79 59
AK032936.1 - 1 - 2 57 59
AK075584.1 - 1 - 2 75 59
AK150895.1 - 1 - 2 64 59
AK151175.1 - 1 - 2 64 59
AK167640.1 - 1 - 2 64 59
AK168009.1 - 1 - 2 67 59
BC024463.1 - 1 - 2 69 59

View Full Product Details