Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Micall1
Entrez Gene ID
27008
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing -like 1
Gene Aliases
260, AI448246, D15Mit260, D15N2e, EST573322, EST820961, MIRab13, mKIAA1668
Chromosome Location
Chr.15: 79108477 - 79136900 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177461.1 NP_803412.1 4 - 5 731 115
GenBank mRNA AK129419.1 - 4 - 5 469 115
AK147587.1 - 4 - 5 732 115
AK147646.1 - 4 - 5 742 115

View Full Product Details