Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Troap
Entrez Gene ID
78733
Gene Name
trophinin associated protein
Gene Aliases
AW476063, E130301L11Rik, tastin
Chromosome Location
Chr.15: 99074973 - 99083409 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162506.1 NP_001155978.1 2 - 3 250 57
NM_030159.1 NP_084435.1 2 - 3 275 57
GenBank mRNA AK021408.1 - 2 - 3 262 57
AK050381.1 - 2 - 3 271 57
AK077509.1 - 2 - 3 240 57
BC080746.1 - 2 - 3 235 57
BC100405.1 - 2 - 3 204 57
BC145227.1 - 2 - 3 648 57

View Full Product Details