Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Troap
Entrez Gene ID
78733
Gene Name
trophinin associated protein
Gene Aliases
AW476063, E130301L11Rik, tastin
Chromosome Location
Chr.15: 99074973 - 99083409 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162506.1 NP_001155978.1 9 - 10 1088 85
NM_030159.1 NP_084435.1 9 - 10 1113 85
GenBank mRNA AK050381.1 - 9 - 10 1109 85
AK077509.1 - 9 - 10 1078 85
BC080746.1 - 9 - 10 1073 85
BC100405.1 - 9 - 10 1042 85
BC145227.1 - 9 - 10 1486 85

View Full Product Details