Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Anks1
Entrez Gene ID
224650
Gene Name
ankyrin repeat and SAM domain containing 1
Gene Aliases
Anks1a, mKIAA0229
Chromosome Location
Chr.17: 27909306 - 28062779 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286040.1 NP_001272969.1 6 - 7 1012 66
NM_001286041.1 NP_001272970.1 5 - 6 949 66
NM_181413.4 NP_852078.1 6 - 7 1012 66
GenBank mRNA AK129094.1 - 5 - 6 883 66
AK147273.1 - 6 - 7 947 66
BC050847.1 - 6 - 7 978 66

View Full Product Details