Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Anks1
Entrez Gene ID
224650
Gene Name
ankyrin repeat and SAM domain containing 1
Gene Aliases
Anks1a, mKIAA0229
Chromosome Location
Chr.17: 27909306 - 28062779 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286040.1 NP_001272969.1 10 - 11 1515 112
NM_001286041.1 NP_001272970.1 9 - 10 1452 112
NM_181413.4 NP_852078.1 10 - 11 1515 112
GenBank mRNA AK129094.1 - 9 - 10 1386 112
AK147273.1 - 10 - 11 1450 112
BC050847.1 - 10 - 11 1481 112

View Full Product Details