Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Srpk1
Entrez Gene ID
20815
Gene Name
serine/arginine-rich protein specific kinase 1
Gene Aliases
AU017960
Chromosome Location
Chr.17: 28589590 - 28622636 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016795.3 NP_058075.2 6 - 7 526 111
GenBank mRNA AB012290.1 - 5 - 6 484 111
AJ224115.1 - 6 - 7 520 111
AK015754.1 - 3 - 4 268 111
AK017187.1 - 6 - 7 523 111
AK141271.1 - 6 - 7 593 111
AK151152.1 - 6 - 7 589 111
AK161346.1 - 6 - 7 504 111
AK161388.1 - 6 - 7 505 111
BC005707.1 - 6 - 7 495 111
BC050761.1 - 6 - 7 526 111

View Full Product Details