Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Jmjd8
Entrez Gene ID
72106
Gene Name
jumonji domain containing 8
Gene Aliases
2610003J06Rik
Chromosome Location
Chr.17: 25829043 - 25831843 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028101.4 NP_082377.2 4 - 5 365 74
GenBank mRNA AK011308.1 - 4 - 5 362 74
AK148079.1 - 4 - 5 343 74
AK155810.1 - 4 - 5 367 74
AK172073.1 - 4 - 5 362 74
BC010800.1 - 4 - 5 327 74

View Full Product Details