Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fbxl16
Entrez Gene ID
214931
Gene Name
F-box and leucine-rich repeat protein 16
Gene Aliases
BC042620, Scirr1
Chromosome Location
Chr.17: 25809050 - 25821265 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164225.1 NP_001157697.1 5 - 6 1609 63
GenBank mRNA BC042620.1 - 4 - 5 1318 63
BC132383.1 - 4 - 5 1405 63
BC137657.1 - 4 - 5 1405 63

View Full Product Details