Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nrk
Entrez Gene ID
27206
Gene Name
Nik related kinase
Gene Aliases
AA407323, AA409510, AI225895, AV168563, AW538185, Nesk
Chromosome Location
Chr.X: 138914312 - 139010532 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013724.2 NP_038752.2 1 - 2 382 121
GenBank mRNA AB020741.1 - 1 - 2 307 121
AB035267.1 - 1 - 2 60 121
AK041377.1 - 1 - 2 382 121
BC068311.1 - 1 - 2 357 121
BC139029.1 - 1 - 2 149 121
BC139031.1 - 1 - 2 149 121

View Full Product Details