The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042673.2 | NP_001036138.1 | 7 - 8 | 1015 | 69 |
NM_001282942.1 | NP_001269871.1 | 6 - 7 | 818 | 69 | |
NM_007985.3 | NP_032011.2 | 7 - 8 | 1015 | 69 | |
GenBank mRNA | AK035488.1 | - | 7 - 8 | 769 | 69 |
AK042226.1 | - | 7 - 8 | 985 | 69 | |
AK080697.1 | - | - | 996 | 69 | |
AK082717.1 | - | 6 - 7 | 796 | 69 | |
AK147025.1 | - | 7 - 8 | 995 | 69 | |
AK163161.1 | - | 7 - 8 | 1016 | 69 | |
BC038609.1 | - | 6 - 7 | 800 | 69 |