Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbln1
Entrez Gene ID
14114
Gene Name
fibulin 1
Gene Aliases
-
Chromosome Location
Chr.15: 85205944 - 85286294 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010180.2 NP_034310.2 2 - 3 307 73
GenBank mRNA AK035388.1 - 2 - 3 260 73
AK083573.1 - 2 - 3 310 73
AK085989.1 - 2 - 3 249 73
AK086451.1 - 2 - 3 308 73
AK137511.1 - 2 - 3 250 73
AK159646.1 - 2 - 3 262 73
AK183901.1 - 2 - 3 191 73
BC007140.1 - 2 - 3 251 73
EU543224.1 - 2 - 3 245 73
X70854.1 - 2 - 3 304 73

View Full Product Details