Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Col1a1
Entrez Gene ID
12842
Gene Name
collagen, type I, alpha 1
Gene Aliases
Col1a-1, Cola-1, Cola1, Mov-13, Mov13
Chromosome Location
Chr.11: 94936270 - 94951867 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007742.3 NP_031768.2 21 - 22 1530 84
GenBank mRNA AK132180.1 - 21 - 22 85
AK158011.1 - 21 - 22 1547 84
AK159283.1 - 21 - 22 1548 84
AK159285.1 - 21 - 22 1550 84
BC042642.1 - 14 - 15 1100 84
BC050014.1 - 21 - 22 1530 84
BC059281.1 - 21 - 22 1540 84
U08020.1 - 21 - 22 1431 84

View Full Product Details