Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr3
Entrez Gene ID
269470
Gene Name
WD repeat domain 3
Gene Aliases
AW546279, D030020G18Rik
Chromosome Location
Chr.3: 100138180 - 100162403 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175552.4 NP_780761.1 2 - 3 262 96
GenBank mRNA AK050738.1 - 2 - 3 230 96
AK050797.1 - 2 - 3 233 96
BC063100.1 - 2 - 3 248 96
BY125127.1 - 2 - 3 263 96

View Full Product Details