Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Rpusd1
- Entrez Gene ID
-
106707
- Gene Name
- RNA pseudouridylate synthase domain containing 1
- Gene Aliases
-
2310051D06Rik, AU019540, Rlucl
- Chromosome Location
-
Chr.17: 25727703 - 25731464 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK009926.1 |
- |
1 - 2 |
115 |
79 |
View Full Product Details