The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025881.3 | NP_080157.1 | 8 - 9 | 927 | 60 |
NM_028190.3 | NP_082466.2 | 8 - 9 | 927 | 60 | |
NR_037905.1 | 9 - 10 | 998 | 60 | ||
NR_037906.1 | 8 - 9 | 1984 | 60 | ||
GenBank mRNA | AK010539.1 | - | 8 - 9 | 879 | 60 |
AK017655.1 | - | 8 - 9 | 887 | 60 | |
BC019194.1 | - | 9 - 10 | 920 | 60 | |
BC034135.1 | - | 7 - 8 | 1093 | 60 | |
BC055875.1 | - | 8 - 9 | 1910 | 60 | |
BQ572015.1 | - | 6 - 7 | 579 | 60 | |
CF951283.1 | - | 4 - 5 | 323 | 60 |