The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252625.1 | NP_001239554.1 | 3 - 4 | 444 | 61 |
NM_029880.3 | NP_084156.2 | 3 - 4 | 387 | 61 | |
GenBank mRNA | AK020666.1 | - | 3 - 4 | 316 | 61 |
AK021033.1 | - | 3 - 4 | 279 | 61 | |
AK036168.1 | - | 3 - 4 | 309 | 61 | |
AK145232.1 | - | 3 - 4 | 335 | 61 | |
AK159187.1 | - | 3 - 4 | 283 | 61 | |
AK159932.1 | - | 3 - 4 | 284 | 61 | |
AK168895.1 | - | 3 - 4 | 278 | 61 | |
BC021466.1 | - | 3 - 4 | 333 | 61 |